Assay Details
Target Gene Details
Entrez Gene ID: | 54353 |
Gene Name: | src family associated phosphoprotein 2 |
Gene Aliases: |
2610021A10Rik, AA960083, BB137539, RA70, SKAP-HOM, Saps, Scap2, mSKAP55R |
Location: |
Chr.6:51859165-52012549 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Skap2 | NM_018773.2 | NP_061243.1 | ||
AB014485.1 | BAA77253.1 | |||
AF051324.1 | AAC99297.1 | |||
AK076000.1 | BAC36111.1 | |||
AK144289.1 | BAE25817.1 | |||
BC003711.1 | AAH03711.1 |
More Information
Set Membership: |
Intragenic Non-exonic |