Assay Details
Target Gene Details
Entrez Gene ID: | 641361 |
Gene Name: | phospholipase A2 inhibitor and LY6/PLAUR domain containing |
Gene Aliases: |
2310033E01Rik |
Location: |
Chr.7:24541654-24546313 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pinlyp | NM_001037143.2 | NP_001032220.1 | ||
AK009589.1 | BAB26378.1 | |||
AK009604.1 | BAB26386.1 | |||
AK009778.1 | BAB26498.1 | |||
AK009924.1 | BAB26587.1 | |||
BC139039.1 | ||||
BC145686.1 |
More Information
Set Membership: |
Intragenic Non-exonic |