Assay Details
Target Gene Details
Entrez Gene ID: | 22044 |
Gene Name: | thyrotropin releasing hormone |
Gene Aliases: |
Pro-TRH, Trf |
Location: |
Chr.6:92242061-92244650 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trh | NM_009426.3 | 2 | 171 | NP_033452.2 |
AK010666.1 | 2 | 170 | ||
BC053493.1 | 2 | 157 | AAH53493.1 | |
CD562178.1 | 2 | 160 | ||
CF951860.1 | 2 | 159 | ||
X59387.1 | 1 | 121 | CAA42030.1 |
More Information
Set Membership: |
Intragenic Exonic |