Assay Details
Target Gene Details
Entrez Gene ID: | 60321 |
Gene Name: | WW domain binding protein 11 |
Gene Aliases: |
2510026P17Rik, D6Wsu113e, Npwbp, SIPP1 |
Location: |
Chr.6:136813654-136828216 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wbp11 | NM_021714.4 | NP_068360.4 | ||
AK011001.1 | ||||
AK031678.1 | BAC27508.1 | |||
AK041079.1 | BAC30812.1 | |||
AK144968.1 | BAE26160.1 | |||
AK145049.1 | ||||
AK145917.1 | BAE26748.1 | |||
AK151989.1 | BAE30854.1 | |||
AK153044.1 | BAE31673.1 | |||
AK153927.1 | BAE32259.1 | |||
AK159257.1 | BAE34938.1 | |||
AK182082.1 | ||||
BC006600.1 | AAH06600.1 | |||
BC021823.1 | AAH21823.1 |