Assay Details
Target Gene Details
Entrez Gene ID: | 27205 |
Gene Name: | podocalyxin-like |
Gene Aliases: |
AW121214, Ly102, PC, PCLP-1, Pclp1 |
Location: |
Chr.6:31519493-31563937 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Podxl | NM_013723.3 | NP_038751.2 | ||
AB028048.1 | BAA86912.1 | |||
AF290209.1 | AAG02458.1 | |||
BC052442.1 | AAH52442.1 | |||
BC054530.1 | AAH54530.1 |
More Information
Set Membership: |
Intragenic Non-exonic |