Assay Details
Target Gene Details
Entrez Gene ID: | 60345 |
Gene Name: | nuclear receptor interacting protein 2 |
Gene Aliases: |
AW491344, NIX1 |
Location: |
Chr.6:128397625-128409784 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrip2 | NM_001162858.1 | NP_001156330.1 | ||
NM_021717.2 | NP_068363.2 | |||
AJ278170.1 | CAB95737.1 | |||
AK134689.1 | BAE22242.1 | |||
AK156526.1 | BAE33744.1 | |||
BC094230.1 | AAH94230.1 | |||
BC115523.1 | ||||
BY718516.1 |
More Information
Set Membership: |
Intragenic Non-exonic |