Assay Details
Target Gene Details
Entrez Gene ID: | 66881 |
Gene Name: | prenylcysteine oxidase 1 |
Gene Aliases: |
1200015P13Rik, AI115532, AI448340, PCL1, Pcly |
Location: |
Chr.6:86386006-86397158 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcyox1 | NM_025823.4 | 5 | 849 | NP_080099.1 |
AK004799.2 | 5 | 804 | BAB23572.1 | |
AK004840.1 | 5 | 804 | BAB23607.1 | |
AK031585.1 | 5 | 805 | BAC27462.1 | |
AK033373.1 | 5 | 805 | BAC28252.1 | |
AK049412.1 | 5 | 802 | BAC33741.1 | |
AK134528.1 | 5 | 850 | BAE22170.1 | |
AK145366.1 | 5 | 809 | BAE26391.1 | |
AK147190.1 | 5 | 817 | BAE27750.1 | |
AK159981.1 | 5 | 805 | BAE35532.1 | |
AK161616.1 | 5 | 805 | BAE36494.1 | |
AK169209.1 | 5 | 805 | BAE40981.1 | |
AK173058.1 | 5 | 771 | BAD32336.1 | |
BC006056.1 | 3 | 460 | AAH06056.1 | |
BC028308.1 | 5 | 819 | AAH28308.1 |
More Information
Set Membership: |
Intragenic Exonic |