Assay Details
Target Gene Details
Entrez Gene ID: | 66881 |
Gene Name: | prenylcysteine oxidase 1 |
Gene Aliases: |
1200015P13Rik, AI115532, AI448340, PCL1, Pcly |
Location: |
Chr.6:86386006-86397158 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcyox1 | NM_025823.4 | NP_080099.1 | ||
AK004799.2 | BAB23572.1 | |||
AK004840.1 | BAB23607.1 | |||
AK031585.1 | BAC27462.1 | |||
AK033373.1 | BAC28252.1 | |||
AK049412.1 | BAC33741.1 | |||
AK134528.1 | BAE22170.1 | |||
AK145366.1 | BAE26391.1 | |||
AK147190.1 | BAE27750.1 | |||
AK159981.1 | BAE35532.1 | |||
AK161616.1 | BAE36494.1 | |||
AK169209.1 | BAE40981.1 | |||
AK173058.1 | BAD32336.1 | |||
BC006056.1 | AAH06056.1 | |||
BC028308.1 | AAH28308.1 |
More Information
Set Membership: |
Intragenic Non-exonic |