Assay Details
Target Gene Details
Entrez Gene ID: | 110109 |
Gene Name: | NOP2 nucleolar protein |
Gene Aliases: |
120kDa, A530002O17, Nol1 |
Location: |
Chr.6:125131883-125144753 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nop2 | NM_138747.2 | NP_620086.2 | ||
AK079914.1 | ||||
AK154006.1 | BAE32311.1 | |||
AK156568.1 | ||||
BC007151.1 | AAH07151.1 | |||
BY244994.1 |