Assay Details
Target Gene Details
Entrez Gene ID: | 114142 |
Gene Name: | forkhead box P2 |
Gene Aliases: |
2810043D05Rik, AI449000, CAG-16, D0Kist7 |
Location: |
Chr.6:14901226-15441977 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxp2 | NM_053242.4 | NP_444472.2 | ||
AK051668.1 | BAC34711.1 | |||
AK080549.1 | ||||
AK082361.1 | BAC38477.1 | |||
AK085080.1 | ||||
AY079003.1 | AAL85482.1 | |||
CN526766.1 |