Assay Details
Target Gene Details
Entrez Gene ID: | 66441 |
Gene Name: | mago-nashi homolog B (Drosophila) |
Gene Aliases: |
2010012C16Rik, Mago, Magoh, Magoh-rs1 |
Location: |
Chr.6:131284388-131293244 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Magohb | NM_025564.2 | NP_079840.2 | ||
AK006303.1 | BAB24516.2 | |||
AK007769.1 | BAB25244.1 | |||
AK008200.1 | BAB25528.1 | |||
AK013310.1 | BAB28784.1 | |||
BC027505.1 | AAH27505.1 | |||
KF051005.1 |