Assay Details
Target Gene Details
Entrez Gene ID: | 56615 |
Gene Name: | microsomal glutathione S-transferase 1 |
Gene Aliases: |
1500002K10Rik, Gst |
Location: |
Chr.6:138140537-138156755 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mgst1 | NM_019946.4 | 2 | 141 | NP_064330.2 |
AF159050.1 | 1 | 40 | AAD51096.1 | |
AK002800.1 | 2 | 142 | BAB22368.1 | |
AK005122.1 | 2 | 142 | BAB23827.1 | |
BC009155.1 | 1 | 80 | AAH09155.1 |
More Information
Set Membership: |
Intragenic Exonic |