Assay Details
Target Gene Details
Entrez Gene ID: | 93695 |
Gene Name: | glycoprotein (transmembrane) nmb |
Gene Aliases: |
DC-HIL, Dchil, ipd |
Location: |
Chr.6:49036518-49058182 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpnmb | NM_053110.4 | NP_444340.3 | ||
AF322054.1 | AAK14240.1 | |||
AJ251685.1 | CAB65272.1 | |||
AK044764.1 | BAC32074.1 | |||
AK076347.1 | BAC36306.1 | |||
AK149740.1 | ||||
AK150739.1 | BAE29813.1 | |||
AK154617.1 | BAE32716.1 | |||
AK159165.1 | BAE34867.1 | |||
AK159270.1 | BAE34949.1 | |||
AK159747.1 | BAE35339.1 | |||
AK159908.1 | ||||
AK170506.1 | BAE41844.1 | |||
AK171347.1 | BAE42404.1 | |||
AK171441.1 | BAE42454.1 | |||
AK171465.1 | BAE42471.1 | |||
AK171474.1 | BAE42479.1 | |||
AK171517.1 | BAE42501.1 | |||
AK171608.1 | BAE42560.1 | |||
AK171618.1 | BAE42567.1 | |||
AK190714.1 | ||||
AK191966.1 | ||||
AK194424.1 | ||||
AK198982.1 | ||||
AK200656.1 | ||||
BC026375.1 | AAH26375.1 |
More Information
Set Membership: |
Intragenic Non-exonic |