Assay Details
Target Gene Details
Entrez Gene ID: | 58991 |
Gene Name: | ghrelin |
Gene Aliases: |
2210006E23Rik, Ghr, MTLRP, MTLRPAP, m46 |
Location: |
Chr.6:113716119-113727485 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ghrl | NM_001286404.1 | 4 | 437 | NP_001273333.1 |
NM_001286405.1 | 3 | 331 | NP_001273334.1 | |
NM_001286406.1 | 3 | 323 | NP_001273335.1 | |
NM_021488.5 | 5 | 488 | NP_067463.2 | |
AB035701.1 | 4 | 430 | BAB19046.1 | |
AJ243503.1 | CAB46500.1 | |||
AK008658.1 | 5 | 458 | BAB25814.1 | |
AK008860.1 | 5 | 458 | BAB25934.1 | |
AY179430.1 | 3 | 321 | AAO27350.1 | |
BC132230.1 | ||||
BC132232.1 |
More Information
Set Membership: |
Intragenic Exonic |