Assay Details
Target Gene Details
Entrez Gene ID: | 27055 |
Gene Name: | FK506 binding protein 9 |
Gene Aliases: |
1810014L23, AW549641, FKBP-63, FKBP-9, FKBP60, FKBP63, FKBP65RS |
Location: |
Chr.6:56832059-56879360 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp9 | NM_012056.2 | 9 | 1564 | NP_036186.2 |
AF090334.1 | 9 | 1490 | AAC72964.1 | |
AF279263.1 | 9 | 1542 | AAF79215.1 | |
AK007499.1 | 7 | 954 | BAB25071.1 | |
AK190681.1 | ||||
AK209013.1 | ||||
AK211009.1 | 2 | 173 | ||
AK212595.1 | ||||
AK218657.1 | 2 | 173 | ||
BC026133.1 | 9 | 1504 | AAH26133.1 | |
BC043129.1 | 9 | 1564 | AAH43129.1 |