Assay Details
Target Gene Details
Entrez Gene ID: | 353172 |
Gene Name: | glycyl-tRNA synthetase |
Gene Aliases: |
GENA202, Gena201, Nmf249, Sgrp23 |
Location: |
Chr.6:55038001-55079504 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gars | NM_180678.3 | 9 | 1116 | NP_851009.2 |
AK012758.2 | 9 | 1111 | ||
AK146238.1 | 9 | 1093 | BAE27003.1 | |
AK165857.1 | 9 | 1064 | BAE38417.1 | |
AK190416.1 | 2 | 180 | ||
AK190448.1 | 3 | 282 | ||
AK191539.1 | 3 | 303 | ||
AK194283.1 | 3 | 258 | ||
AK195104.1 | 3 | 263 | ||
AK196051.1 | 3 | 282 | ||
AK203488.1 | 3 | 241 | ||
AK212952.1 | 3 | 301 | ||
BC021747.1 | 9 | 1097 | AAH21747.1 |
More Information
Set Membership: |
Intragenic Exonic |