Assay Details
Target Gene Details
Entrez Gene ID: | 353172 |
Gene Name: | glycyl-tRNA synthetase |
Gene Aliases: |
GENA202, Gena201, Nmf249, Sgrp23 |
Location: |
Chr.6:55038001-55079504 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gars | NM_180678.3 | 12 | 1545 | NP_851009.2 |
AK012758.2 | 12 | 1540 | ||
AK146238.1 | 12 | 1522 | BAE27003.1 | |
AK165857.1 | 12 | 1493 | BAE38417.1 | |
AK192105.1 | 1 | 131 | ||
AK201644.1 | 1 | 56 | ||
AK204968.1 | 1 | 105 | ||
AK207297.1 | 1 | 56 | ||
AK215449.1 | 2 | 145 | ||
BC021747.1 | 12 | 1526 | AAH21747.1 |
More Information
Set Membership: |
Intragenic Exonic |