Assay Details
Target Gene Details
Entrez Gene ID: | 353172 |
Gene Name: | glycyl-tRNA synthetase |
Gene Aliases: |
GENA202, Gena201, Nmf249, Sgrp23 |
Location: |
Chr.6:55038001-55079504 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gars | NM_180678.3 | 1 | 75 | NP_851009.2 |
AK012758.2 | 1 | 70 | ||
AK146238.1 | 1 | 52 | BAE27003.1 | |
AK165857.1 | BAE38417.1 | |||
BC021747.1 | 1 | 56 | AAH21747.1 | |
BY309346.1 | 1 | 75 |
More Information
Set Membership: |
Intragenic Exonic |