Assay Details
Target Gene Details
Entrez Gene ID: | 53621 |
Gene Name: | CCR4-NOT transcription complex, subunit 4 |
Gene Aliases: |
Not4, Not4h, Not4hp |
Location: |
Chr.6:35022065-35133737 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cnot4 | NM_001164411.1 | 9 | 1282 | NP_001157883.1 |
NM_001164412.1 | 9 | 1282 | NP_001157884.1 | |
NM_001164413.1 | 9 | 1282 | NP_001157885.1 | |
NM_001164414.1 | 9 | 1291 | NP_001157886.1 | |
NM_016877.4 | 9 | 1291 | NP_058573.3 | |
AK028190.1 | 9 | 1152 | BAC25801.1 | |
AK029753.1 | 9 | 1260 | ||
AK032248.1 | 9 | 1271 | BAC27779.1 | |
AK075887.1 | 9 | 1272 | ||
AK083811.1 | 1 | 3403 | ||
AK144760.1 | 9 | 1249 | ||
AK165997.1 | 9 | 1268 | BAE38508.1 | |
BC048243.1 | 9 | 1170 | ||
BC058778.1 | 9 | 1234 | AAH58778.1 | |
U71269.1 | 9 | 1277 | AAD00181.1 |
More Information
Set Membership: |
Intragenic Exonic |