Assay Details
Target Gene Details
Entrez Gene ID: | 68396 |
Gene Name: | N-acetyltransferase 8 (GCN5-related, putative) |
Gene Aliases: |
0610037O16Rik, CCNAT, Cml4 |
Location: |
Chr.6:85830387-85836636 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nat8 | NM_023455.3 | NP_075944.1 | ||
AF163317.1 | AAF80486.1 | |||
AK002784.1 | BAB22356.1 | |||
BB846150.1 | ||||
BC019517.1 | AAH19517.1 | |||
BY081766.1 |
More Information
Set Membership: |
Intragenic Non-exonic |