Assay Details
Target Gene Details
Entrez Gene ID: | 12638 |
Gene Name: | cystic fibrosis transmembrane conductance regulator |
Gene Aliases: |
AW495489, Abcc7 |
Location: |
Chr.6:18170262-18322774 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cftr | NM_021050.2 | 10 | 1427 | NP_066388.1 |
AK033621.1 | 10 | 1345 | BAC28395.1 | |
M60493.1 | 10 | 1428 | AAA18903.1 | |
M69298.1 | 10 | 1426 | AAA37417.1 | |
M84613.1 | 1 | 81 |
More Information
Set Membership: |
Intragenic Exonic |