Assay Details
Target Gene Details
Entrez Gene ID: | 65106 |
Gene Name: | ADP-ribosylation factor-like 6 interacting protein 5 |
Gene Aliases: |
5930404D22Rik, AV001879, Aip-5, Gtrap3-18, addiscin |
Location: |
Chr.6:97210792-97233315 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arl6ip5 | NM_022992.2 | 2 | 267 | NP_075368.1 |
AF133912.1 | 2 | 263 | AAD33050.1 | |
AF265214.1 | 2 | 238 | AAL74056.1 | |
AF421860.1 | 2 | 273 | AAL77876.1 | |
AK005259.1 | 2 | 264 | BAB23912.1 | |
AK008519.1 | 2 | 247 | BAB25717.1 | |
AK076519.1 | 2 | 264 | BAC36376.1 | |
AK077826.1 | 2 | 274 | ||
AK088205.1 | 2 | 262 | BAC40209.1 | |
BC003897.1 | 2 | 267 | AAH03897.1 | |
D87211.1 | 2 | 310 | BAC24103.1 |
More Information
Set Membership: |
Intragenic Exonic |