Assay Details
Target Gene Details
Entrez Gene ID: | 12468 |
Gene Name: | chaperonin containing Tcp1, subunit 7 (eta) |
Gene Aliases: |
AA408524, AL022769, Ccth, Cctz |
Location: |
Chr.6:85451505-85468477 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cct7 | NM_007638.4 | NP_031664.3 | ||
AK012201.1 | ||||
AK077781.1 | BAC37005.1 | |||
AK146902.1 | BAE27516.1 | |||
AK150160.1 | BAE29350.1 | |||
AK166888.1 | BAE39094.1 | |||
AK167367.1 | BAE39463.1 | |||
AK167827.1 | BAE39849.1 | |||
AK167938.1 | BAE39942.1 | |||
AK168087.1 | BAE40060.1 | |||
AK168272.1 | BAE40218.1 | |||
AK169426.1 | BAE41168.1 | |||
AK196078.1 | ||||
BC008255.1 | AAH08255.1 | |||
Z31399.1 | CAA83274.1 |
More Information
Set Membership: |
Intragenic Non-exonic |