Assay Details
Target Gene Details
Entrez Gene ID: | 109880 |
Gene Name: | Braf transforming gene |
Gene Aliases: |
9930012E13Rik, AA120551, AA387315, AA473386, B-raf, Braf-2, Braf2, C230098H17, C87398, D6Ertd631e |
Location: |
Chr.6:39603231-39725656 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Braf | NM_139294.5 | NP_647455.3 | ||
AF454556.1 | AAQ04690.1 | |||
AF454557.1 | AAQ04691.1 | |||
AF454558.1 | AAQ04692.1 | |||
AF454559.1 | AAQ04693.1 | |||
AK140431.1 | BAE24384.1 | |||
AK164972.1 | 2 | 4017 |
More Information
Set Membership: |
Intragenic Non-exonic |