Assay Details
Target Gene Details
Entrez Gene ID: | 20346 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A |
Gene Aliases: |
Hsema-I, SEMA1, SemD, Semad, coll-1 |
Location: |
Chr.5:13125371-13606234 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sema3a | NM_001243072.1 | 6 | 875 | NP_001230001.1 |
NM_001243073.1 | 6 | 715 | NP_001230002.1 | |
NM_009152.4 | 5 | 1143 | NP_033178.2 | |
AK140817.1 | 6 | 716 | BAE24487.1 | |
AK164242.1 | 8 | 918 | BAE37700.1 | |
BC057588.1 | 5 | 790 | AAH57588.1 | |
BC090844.1 | 6 | 875 | AAH90844.1 | |
D85028.2 | 5 | 1145 | BAA19773.1 | |
L40484.1 | 2 | 176 | AAA73934.1 | |
L41541.1 | 5 | 494 | AAL77611.1 | |
X85993.1 | 5 | 604 | CAA59985.1 |
More Information
Set Membership: |
Intragenic Exonic |