Assay Details
Target Gene Details
Entrez Gene ID: | 22223 |
Gene Name: | ubiquitin carboxy-terminal hydrolase L1 |
Gene Aliases: |
AW822034, C88048, PGP 9.5, PGP9.5, R75593, UCH-L1, UCHL-1, gad |
Location: |
Chr.5:66676121-66687234 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uchl1 | NM_011670.2 | 1 | 62 | NP_035800.2 |
CX241178.1 | 1 | 62 |
Target Gene Details
Entrez Gene ID: | 101202684 |
Gene Name: | ubiquitin carboxy-terminal hydrolase L1, opposite strand |
Gene Aliases: |
Gm16832, Uchl1as |
Location: |
Chr.5:66626495-66676497 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uchl1os | NR_102714.1 | 1 | 316 | |
AK078321.1 | 1 | 317 |
More Information
Set Membership: |
Intragenic Exonic |