Assay Details
Target Gene Details
Entrez Gene ID: | 100763 |
Gene Name: | ubiquitin protein ligase E3C |
Gene Aliases: |
AI853514, mKIAA0010 |
Location: |
Chr.5:29569242-29676077 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube3c | NM_133907.3 | 17 | 2435 | NP_598668.1 |
AK029973.1 | 17 | 2245 | BAC26709.1 | |
AK046056.1 | 17 | 2436 | BAC32585.1 | |
AK122187.1 | 17 | 2370 | BAC65469.1 | |
BC021525.1 | 6 | 680 | AAH21525.1 | |
BC120731.1 | 17 | 2182 | ||
BC137626.1 | 17 | 2182 |
More Information
Set Membership: |
Intragenic Exonic |