Assay Details
Target Gene Details
Entrez Gene ID: | 100763 |
Gene Name: | ubiquitin protein ligase E3C |
Gene Aliases: |
AI853514, mKIAA0010 |
Location: |
Chr.5:29569242-29676077 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube3c | NM_133907.3 | 13 | 1962 | NP_598668.1 |
AK029973.1 | 13 | 1772 | BAC26709.1 | |
AK045211.1 | 13 | 1760 | ||
AK046056.1 | 13 | 1963 | BAC32585.1 | |
AK122187.1 | 13 | 1897 | BAC65469.1 | |
BC021525.1 | 2 | 207 | AAH21525.1 | |
BC120731.1 | 13 | 1709 | ||
BC137626.1 | 13 | 1709 |
More Information
Set Membership: |
Intragenic Exonic |