Assay Details
Target Gene Details
Entrez Gene ID: | 69982 |
Gene Name: | serine peptidase inhibitor, Kazal type 2 |
Gene Aliases: |
1700007F22Rik, AV038945, HUSI-II |
Location: |
Chr.5:77205106-77216260 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spink2 | NM_001289764.1 | NP_001276693.1 | ||
NM_001289767.1 | NP_001276696.1 | |||
NM_001289768.1 | NP_001276697.1 | |||
NM_183284.3 | NP_899107.1 | |||
AK005709.1 | BAC25122.1 | |||
BC087876.1 | AAH87876.1 | |||
DV068802.1 |
More Information
Set Membership: |
Intragenic Non-exonic |