Assay Details
Target Gene Details
Entrez Gene ID: | 67956 |
Gene Name: | SET domain containing (lysine methyltransferase) 8 |
Gene Aliases: |
2410195B05Rik, AA617402, AW536475, PR-SET7 |
Location: |
Chr.5:124439930-124462308 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Setd8 | NM_030241.2 | NP_084517.2 | ||
AK010593.1 | BAC25304.1 | |||
AK010835.1 | ||||
AK030904.1 | BAC27178.1 | |||
AK088097.1 | ||||
BB619898.1 | ||||
BC108333.1 | AAI08334.1 | |||
BY041630.1 |
More Information
Set Membership: |
Intragenic Non-exonic |