Assay Details
Target Gene Details
Entrez Gene ID: | 330222 |
Gene Name: | sidekick homolog 1 (chicken) |
Gene Aliases: |
5330440E10, 6720466O15Rik |
Location: |
Chr.5:141241497-142215586 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sdk1 | NM_177879.5 | 11 | 1854 | NP_808547.3 |
AK147578.1 | 11 | 1899 | BAE28004.1 | |
AY353236.1 | 7 | 1004 | AAQ57662.1 | |
BC060237.1 | 11 | 1438 | AAH60237.1 | |
BC138453.1 | 7 | 875 |