Assay Details
Target Gene Details
Entrez Gene ID: | 68364 |
Gene Name: | RIKEN cDNA 0610030E20 gene |
Gene Aliases: |
1500031N04Rik, 2810411C16Rik, AI461894 |
Location: |
Chr.6:72347023-72353162 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
0610030E20Rik | NM_026696.1 | NP_080972.1 | ||
AK002701.1 | BAB22295.1 | |||
AK085662.1 | 1 | 1333 | ||
BC064080.1 | ||||
BC066053.1 | ||||
BC117812.1 |