Assay Details
Target Gene Details
Entrez Gene ID: | 20492 |
Gene Name: | stem-loop binding protein |
Gene Aliases: |
- |
Location: |
Chr.5:33640055-33652574 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slbp | NM_001289724.1 | NP_001276653.1 | ||
NM_001289725.1 | NP_001276654.1 | |||
NM_009193.2 | NP_033219.1 | |||
AK016826.1 | ||||
AK154053.1 | BAE32343.1 | |||
AK162121.1 | BAE36737.1 | |||
AK210098.1 | ||||
BC029631.1 | AAH29631.1 | |||
U75680.1 | AAC53530.1 |
More Information
Set Membership: |
Intragenic Non-exonic |