Assay Details
Target Gene Details
Entrez Gene ID: | 212996 |
Gene Name: | Williams-Beuren syndrome chromosome region 17 homolog (human) |
Gene Aliases: |
AA388321, E330012B09Rik |
Location: |
Chr.5:130874951-131307522 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wbscr17 | NM_145218.3 | 5 | 1743 | NP_660253.2 |
AB175684.1 | 5 | 821 | BAD91810.1 | |
AF467979.1 | 5 | 1024 | AAM62404.1 | |
AK035817.1 | 5 | 1497 | BAC29197.1 | |
AK048758.1 | 5 | 1488 | BAC33446.2 | |
AK051281.1 | 5 | 2143 | BAC34591.2 | |
BC158110.1 | 5 | 1770 |
More Information
Set Membership: |
Intragenic Exonic |