Assay Details
Target Gene Details
Entrez Gene ID: | 243084 |
Gene Name: | transmembrane protease, serine 11e |
Gene Aliases: |
A030012E10, AY787860, DESC1, TTSP |
Location: |
Chr.5:86705186-86745807 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmprss11e | NM_172880.2 | NP_766468.1 | ||
AK037235.1 | BAC29770.1 | |||
AK162701.1 | ||||
AY787860.1 | AAV52922.1 | |||
BC115432.1 | ||||
BC115433.1 |
More Information
Set Membership: |
Intragenic Non-exonic |