Assay Details
Target Gene Details
Entrez Gene ID: | 20349 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E |
Gene Aliases: |
6430702L12, AA408817, Semah, mKIAA0331 |
Location: |
Chr.5:14025276-14256689 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sema3e | NM_011348.2 | 14 | 2254 | NP_035478.2 |
AF034744.1 | 14 | 1989 | AAD01996.1 | |
AK049580.1 | 1 | 991 | ||
AK129116.1 | 9 | 1019 | BAC97926.1 | |
AK141295.1 | 13 | 2977 | ||
AK142340.1 | 14 | 2081 | BAE25038.1 | |
AK159214.1 | 14 | 2045 | BAE34903.1 | |
BC057956.1 | 14 | 2152 | AAH57956.1 | |
Z80941.1 | 14 | 1620 | CAB02590.1 | |
Z93947.1 | 14 | 2232 | CAB07987.1 | |
Z93948.1 | 14 | 2232 | CAB07988.1 |
More Information
Set Membership: |
Intragenic Exonic |