Assay Details
Target Gene Details
Entrez Gene ID: | 435845 |
Gene Name: | transmembrane protease, serine 11c |
Gene Aliases: |
Tmprss13 |
Location: |
Chr.5:86231481-86289308 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmprss11c | NM_001030297.2 | NP_001025468.1 | ||
AM260217.1 | ||||
BC147157.1 | ||||
BC147158.1 |
More Information
Set Membership: |
Intragenic Non-exonic |