Assay Details
Target Gene Details
Entrez Gene ID: | 108151 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3D |
Gene Aliases: |
4631426B19Rik |
Location: |
Chr.5:12383166-12588943 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sema3d | NM_028882.4 | 2 | 649 | NP_083158.3 |
AK028900.1 | 2 | 431 | BAC26185.1 | |
AK031704.1 | 2 | 261 | BAC27522.1 | |
AK051165.1 | 2 | 310 | BAC34542.1 | |
AK052671.1 | 2 | 650 | BAC35092.1 | |
AK087013.1 | 2 | 251 | BAC39783.2 | |
AK134412.1 | 2 | 285 | BAE22134.1 | |
AK153839.1 | 2 | 310 | ||
BC138131.1 | 2 | 192 |
More Information
Set Membership: |
Intragenic Exonic |