Assay Details
Target Gene Details
Entrez Gene ID: | 50791 |
Gene Name: | membrane associated guanylate kinase, WW and PDZ domain containing 2 |
Gene Aliases: |
AIP-1, Acvri1, Acvrinp1, Acvrip1, Magi-2, S-SCAM, mKIAA0705 |
Location: |
Chr.5:19223283-20704792 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Magi2 | NM_001170745.1 | 3 | 758 | NP_001164216.1 |
NM_001170746.1 | 4 | 923 | NP_001164217.1 | |
NM_015823.3 | 3 | 758 | NP_056638.1 | |
AK122349.1 | 2 | 218 | BAC65631.3 | |
AK147530.1 | 3 | 759 | BAE27976.1 | |
BC059005.1 | 3 | 415 | AAH59005.1 |