Assay Details
Target Gene Details
Entrez Gene ID: | 56361 |
Gene Name: | pseudouridine synthase 1 |
Gene Aliases: |
A730013B20Rik, MPUS1, mPus1p |
Location: |
Chr.5:110773667-110781056 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pus1 | NM_001025561.3 | NP_001020732.1 | ||
NM_001025562.2 | NP_001020733.1 | |||
NM_019700.4 | NP_062674.2 | |||
AB041563.1 | BAA95047.1 | |||
AF116237.1 | AAD21041.1 | |||
AK089258.1 | BAC40817.1 | |||
AK138570.1 | BAE23701.1 | |||
AK161521.1 | BAE36440.1 | |||
BC021446.1 | ||||
BC034359.1 | ||||
BU515425.1 | ||||
CB059120.1 |
More Information
Set Membership: |
Intragenic Non-exonic |