Assay Details
Target Gene Details
Entrez Gene ID: | 57349 |
Gene Name: | pro-platelet basic protein |
Gene Aliases: |
2400003M24Rik, AI854500, CTAP3, CTAPIII, Cxcl7, LA-PF4, LDGF, MDGF, NAP-2, NAP-2-L1, Scyb7, TGB, TGB1, THBGB1, b-TG1, beta-TG |
Location: |
Chr.5:90768518-90770060 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppbp | NM_023785.2 | NP_076274.1 | ||
AB042817.1 | BAB83941.1 | |||
AF219112.1 | AAG36786.1 | |||
AF278700.1 | AAK69476.1 | |||
AK010273.1 | BAB26811.1 | |||
AK157077.1 | BAE33955.1 | |||
AK157141.1 | BAE33977.1 | |||
AK158061.1 | BAE34343.1 | |||
AK171720.1 | BAE42631.1 | |||
BC127043.1 | ||||
BC127044.1 |
More Information
Set Membership: |
Intragenic Non-exonic |