Assay Details
Target Gene Details
Entrez Gene ID: | 26433 |
Gene Name: | procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3 |
Gene Aliases: |
AI414586, LH3 |
Location: |
Chr.5:136987019-136996646 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plod3 | NM_011962.3 | NP_036092.1 | ||
AF046783.1 | AAD54618.1 | |||
AK013195.1 | BAB28704.1 | |||
AK033360.1 | BAC28246.1 | |||
AK053320.1 | ||||
AK088948.1 | BAC40669.1 | |||
AK157741.1 | ||||
AK204070.1 | ||||
BC043047.1 | AAH43047.1 | |||
BC054734.1 | AAH54734.1 |
More Information
Set Membership: |
Intragenic Non-exonic |