Assay Details
Target Gene Details
Entrez Gene ID: | 19699 |
Gene Name: | reelin |
Gene Aliases: |
reeler, rl |
Location: |
Chr.5:21884454-22344705 on Build GRCm38 |
Assay Gene Location: | Within Exon 62 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Reln | NM_011261.2 | 62 | 10343 | NP_035391.2 |
AK017094.1 | 7 | 916 | BAB30592.1 | |
BC118041.1 | 16 | 2492 | ||
D63520.1 | 21 | 3593 | BAA09788.1 | |
U24703.1 | 62 | 10328 | AAB91599.1 |
More Information
Set Membership: |
Intragenic Exonic |