Assay Details
Target Gene Details
Entrez Gene ID: | 100316903 |
Gene Name: | predicted gene 20605 |
Gene Aliases: |
Lrch4-sap25 |
Location: |
Chr.5:137629123-137642896 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm20605 | NR_033148.1 |
Target Gene Details
Entrez Gene ID: | 231798 |
Gene Name: | leucine-rich repeats and calponin homology (CH) domain containing 4 |
Gene Aliases: |
2810008P14Rik, 2900069C24Rik, AI558103, LRN, LRRN4, SAP25, mFLJ00248 |
Location: |
Chr.5:137629123-137641099 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrch4 | NM_001168652.1 | NP_001162123.1 | ||
NM_146164.2 | NP_666276.1 | |||
AK088331.1 | BAC40287.1 | |||
BC012525.1 | AAH12525.1 | |||
CF729493.1 | ||||
DV661712.1 |