Assay Details
Target Gene Details
Entrez Gene ID: | 19699 |
Gene Name: | reelin |
Gene Aliases: |
reeler, rl |
Location: |
Chr.5:21884454-22344705 on Build GRCm38 |
Assay Gene Location: | Within Exon 48 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Reln | NM_011261.2 | 48 | 7843 | NP_035391.2 |
AK047608.1 | 2 | 74 | ||
BC118041.1 | ||||
D63520.1 | 7 | 1093 | BAA09788.1 | |
U24703.1 | 48 | 7828 | AAB91599.1 |
More Information
Set Membership: |
Intragenic Exonic |