Assay Details
Target Gene Details
Entrez Gene ID: | 17527 |
Gene Name: | MpV17 mitochondrial inner membrane protein |
Gene Aliases: |
Tg.Mpv17 |
Location: |
Chr.5:31140663-31154532 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpv17 | NM_001294322.1 | NP_001281251.1 | ||
NM_001294324.1 | NP_001281253.1 | |||
NM_008622.6 | NP_032648.1 | |||
AK040477.1 | ||||
BC013452.1 | AAH13452.1 | |||
BY258057.1 | ||||
CK622051.1 | ||||
DV062332.1 | ||||
DV660230.1 | ||||
M36411.1 | AAA39736.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |