Assay Details
Target Gene Details
Entrez Gene ID: | 53324 |
Gene Name: | neuronal pentraxin 2 |
Gene Aliases: |
Narp, np2 |
Location: |
Chr.5:144545887-144557478 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nptx2 | NM_016789.3 | 3 | 896 | NP_058069.1 |
AF049124.1 | 3 | 786 | AAC05131.1 | |
AK134806.1 | 3 | 897 | BAE22293.1 | |
BC026054.1 | 3 | 891 | AAH26054.1 | |
DV649643.1 | 3 | 335 |