Assay Details
Target Gene Details
Entrez Gene ID: | 108099 |
Gene Name: | protein kinase, AMP-activated, gamma 2 non-catalytic subunit |
Gene Aliases: |
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS |
Location: |
Chr.5:24862735-25101906 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prkag2 | NM_145401.2 | 4 | 1050 | NP_663376.2 |
AK032238.1 | 2 | 559 | BAC27775.1 | |
AK154296.1 | 4 | 1051 | BAE32497.1 | |
BC015283.1 | 4 | 838 | AAH15283.1 |
More Information
Set Membership: |
Intragenic Exonic |