Assay Details
Target Gene Details
Entrez Gene ID: | 76421 |
Gene Name: | RIKEN cDNA 1700028K03 gene |
Gene Aliases: |
- |
Location: |
Chr.5:107534711-107551542 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700028K03Rik | NM_175241.1 | NP_780450.1 | ||
AK032886.1 | ||||
CN835533.1 |
Target Gene Details
Entrez Gene ID: | 170823 |
Gene Name: | glomulin, FKBP associated protein |
Gene Aliases: |
9330160J16Rik, AW227515, Fap48, Fap68 |
Location: |
Chr.5:107548967-107597888 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glmn | NM_001161738.1 | 19 | 1861 | NP_001155210.1 |
NM_001161739.1 | 17 | 1683 | NP_001155211.1 | |
NM_133248.2 | 19 | 1875 | NP_573511.2 | |
AJ566083.1 | 18 | 1743 | CAD92739.1 | |
AK034160.1 | 19 | 1862 | BAC28612.1 | |
AK165661.1 | 19 | 1846 | BAE38323.1 | |
AK169045.1 | 19 | 1847 | BAE40833.1 | |
AK172659.1 | 17 | 1621 | BAE43119.1 | |
BC003446.1 | 19 | 1811 | AAH03446.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |