Assay Details
Target Gene Details
Entrez Gene ID: | 80752 |
Gene Name: | family with sequence similarity 20, member C |
Gene Aliases: |
C76981, DMP-4, DMP4, GEF-CK, mKIAA4081 |
Location: |
Chr.5:138754306-138810072 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam20c | NM_030565.6 | 2 | 1269 | NP_085042.2 |
AK141453.1 | 1 | 2839 | ||
AK149780.1 | 3 | 1349 | ||
AK154810.1 | 2 | 1265 | ||
AK220286.1 | 2 | 786 | BAD90211.1 | |
AY778962.1 | 2 | 1250 | AAV91326.1 | |
BC025814.1 | 2 | 1220 | AAH25814.1 | |
BC025826.1 | 2 | 1250 | AAH25826.1 |